missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ ProShield™ 50 Coverall (all)

Catalog No. 19065500E
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
Size (English) Chest:
37.25 to 40.75 in.
40.75 to 44.25
44.25 to 47.75 in.
Height (English):
5.22 to 6.16 ft.
5.41 to 5.75 ft.
6 to 6.33 ft.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Size (English) Chest Height (English)
19065500E 2X-Large 44.25 to 47.75 in. 6 to 6.33 ft.
19065500C Large 37.25 to 40.75 in. 5.41 to 5.75 ft.
19065500D X-Large 40.75 to 44.25 5.22 to 6.16 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Should not be used around heat, flame, sparks or potentially flammable or explosive environments.
Catalog No. 19065500E Supplier DuPont™ Supplier No. NB120SWH2X002500
Only null left
Should not be used around heat, flame, sparks or potentially flammable or explosive environments.

Microporous film coveralls provide protection from non-hazardous aerosols, light liquid splash, and dry particles.

  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Manufactured under specifications that do not contain natural rubber latex

Specifications

Product Type Single-use Coveralls
Size 2X-Large
Gender Unisex
Material ProShield
Color White
Disposable Single-use
Ankle Style Open
Cuff Style Open
Neck Style Collared
Hood Type Unhooded
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Collar Type Laydown
Model NB120S
Size (English) Chest 44.25 to 47.75 in.
Size (English) Inseam 28.75 in.
Size (Metric) Inseam 73.02 cm
Thickness 10 mil
For Use With (Application) Janitorial, Sanitation, General Industrial, Maintenance
Length (English) Sleeve 36.25 in.
Length (Metric) Sleeve 92.08 cm
Width (English) Chest 28.75 in.
Width (Metric) Chest 73.02 cm
Height (English) 6 to 6.33 ft.
Height (Metric) 1.83 to 1.93 m
Quantity 25/Cs.
Show More Show Less