missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ ProShield™ 10 Hooded Coverall
Spunbond meltblown spunbond (SMS) coveralls protect against non-hazardous particles and light liquid splashes in a variety of industries.
Supplier: DuPont™ PB127SGY4X002500
Description
- Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
- Attached hood with elastic around face opening.
- Elastic opening for tighter fit at wrist
- Elastic opening for tighter fit at ankle
- Vend packed
- Manufactured under specifications that do not contain natural rubber latex
Specifications
| General Purpose Coveralls | |
| Unisex | |
| Gray | |
| Elastic | |
| Hooded | |
| Front Zipper | |
| Non-sterile | |
| Yes | |
| None | |
| 25/Cs. |
| 4X-Large | |
| ProShield | |
| Single-use | |
| Elastic | |
| Attached | |
| Serged | |
| Particle | |
| PB127SWH | |
| General Maintenance, Janitorial, Cleaning |