missing translation for 'onlineSavingsMsg'
Learn More

Contec™ PROSAT™ Sterile Presaturated Nonwoven Wipes
GREENER_CHOICE

Catalog No. 191500710
Change view
Click to view available options
Dimensions (L x W):
22.86 x 22.86 cm (9 x 9 in.)
27.9 x 23 cm (11 x 9 in.)
Packaging Quantity:
24 Pk.
40 Packs of 20 Wipes
50 Pk.
Saturation Solution:
70% IPA, 30% DI Water
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Packaging Quantity Saturation Solution
191500710 27.9 x 23 cm (11 x 9 in.) 24 Pk. 70% IPA, 30% DI Water
191500706 27.9 x 23 cm (11 x 9 in.) 50 Pk. 70% IPA, 30% DI Water
19170356 22.86 x 22.86 cm (9 x 9 in.) 40 Packs of 20 Wipes 70% IPA, 30% DI Water
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 191500710 Supplier Contec™ Supplier No. PSC20001
Only null left

Wipe down isolators, cabinets, hoods, and other cleanroom equipment and tools. Contec™ PROSAT™ Sterile Presaturated Nonwoven Wipes are ideal for cleanup following BD RODAC™ plate sampling. These wipes are designed specifically for sterile environments.

  • Designed for pharmaceutical, biotech, and medical device applications
  • Choose spunlaced nonwoven 100% polyester wipes for improved cleanliness, economical hydroentangled polyester/cellulose wipers, or meltblown polypropylene wipes for improved durability
  • Conveniently presaturated with 70/30 blend of semi-conductor grade IPA and DI water to reduce VOC emissions and hazardous waste
  • All wipes are sterilized by gamma radiation to a 10-6 Sterility Assurance Level and validated per AAMI ISO 11137 (Method 1) guidelines
  • Certificate of Sterility and Certificate of Irradiation included
  • Environment: ISO 5-8, Grade A/B
  • Individually lot numbered resealable pouches with tamper-evident labels ensure product integrity and cleanliness

Compliance

Validated sterile to a Sterility Assurance Level of 10-6 in accordance with AAMI/ISO 1137 Method 1 Guidelines.

Specifications

Product Type Pre-saturated Wipe
Length (English) 9 in.
Length (Metric) 23 cm
Width (English) 11 in.
Width (Metric) 27.9 cm
Material Polyester Cellulose
Certifications/Compliance ISO Class 5-8
Dimensions (L x W) 27.9 x 23 cm (11 x 9 in.)
For Use With (Application) Ideal for routine cleaning and wipe down of lab tools, instruments and other equipment and designed for sterile environments
Packaging Quantity 24 Pk.
Saturation Solution 70% IPA, 30% DI Water
Sterility Sterile
Show More Show Less