Learn More
Contec™ PROSAT™ Sterile Presaturated Nonwoven Wipes

Wipe down isolators, cabinets, hoods, and other cleanroom equipment and tools. Contec™ PROSAT™ Sterile Presaturated Nonwoven Wipes are ideal for cleanup following BD RODAC™ plate sampling. These wipes are designed specifically for sterile environments.
Supplier: Contec™ PSC20005
Description
Wipe down isolators, cabinets, hoods, and other cleanroom equipment and tools. Contec™ PROSAT™ Sterile Presaturated Nonwoven Wipes are ideal for cleanup following BD RODAC™ plate sampling. These wipes are designed specifically for sterile environments.
- Designed for pharmaceutical, biotech, and medical device applications
- Choose spunlaced nonwoven 100% polyester wipes for improved cleanliness, economical hydroentangled polyester/cellulose wipers, or meltblown polypropylene wipes for improved durability
- Conveniently presaturated with 70/30 blend of semi-conductor grade IPA and DI water to reduce VOC emissions and hazardous waste
- All wipes are sterilized by gamma radiation to a 10-6 Sterility Assurance Level and validated per AAMI ISO 11137 (Method 1) guidelines
- Certificate of Sterility and Certificate of Irradiation included
- Environment: ISO 5-8, Grade A/B
- Individually lot numbered resealable pouches with tamper-evident labels ensure product integrity and cleanliness
Compliance
Validated sterile to a Sterility Assurance Level of 10-6 in accordance with AAMI/ISO 1137 Method 1 Guidelines.
Chemical Identifiers
| Pre-saturated Wipe | |
| Sterile |
Specifications
| Pre-saturated Wipe | |
| 23 cm | |
| 23 cm | |
| ISO Class 5-8 | |
| Ideal for routine cleaning and wipe down of lab tools, instruments and other equipment and designed for sterile environments | |
| 70% IPA, 30% DI Water |
| 9 in. | |
| 9 in. | |
| Polyester Cellulose | |
| 27.9 x 23 cm (11 x 9 in.) | |
| 50 Pk. | |
| Sterile |