missing translation for 'onlineSavingsMsg'
Learn More

Propyne (ca. 3-4% in Heptane), TCI America™

Supplier:  TCI America P1881200G

Catalog No. P1881200G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Propyne (ca. 3-4% in Heptane)
Colorless
MFCD00036235
1206
MWWATHDPGQKSAR-UHFFFAOYSA-N
prop-1-yne
6335
40.07
74-99-7
C3H4
200 g
Methylacetylene
CC#C
40.065
CHEBI:48086
Liquid

Chemical Identifiers

74-99-7
40.065
MWWATHDPGQKSAR-UHFFFAOYSA-N
6335
prop-1-yne
C3H4
MFCD00036235
Methylacetylene
CHEBI:48086
CC#C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0015

RTECSNumber : UK4250000

TSCA : Yes

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions