missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Propylphosphonic anhydride solution

Catalog No. 431303500ML
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
431303500ML 500 mL
431303100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 431303500ML Supplier Sigma Aldrich Supplier No. 431303500ML
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Propylphosphonic anhydride (t3p) is a reactive n-propyl phosphonic acid cyclic anhydride. it is a mild and low toxic coupling agent used in peptide synthesis. t3p also acts as a promoter and water scavenger in the Friedländer annulation reaction. it participates in the conversion of carboxylic acids and amides into nitriles, formation of weinreb amides, ester synthesis, dehydrations, oxidation of alcohols, isonitrile synthesis, synthesis of alkenes from alcohols and c-c coupling reactions. t3p delivers outstanding advantages over traditional reagents, such as broad functional group tolerance, low epimerization, and water-soluble by-products and hence gives high purity and yield of the product.propylphosphonic anhydride may be used in the following studies:
  • As coupling agent for the synthesis of bispyridine-based ligands, which are used as bridging linkers in multinuclear platinum anticancer drugs
  • Microwave-assissted fischer indolization of arylhydrazines
  • As acid activating agent for the direct synthesis of acid azides from carboxylic acids
  • One-pot synthesis of coumarins
  • Microwave-mediated synthesis of carbocyclic and heterocyclic fused quinolones
  • One-pot synthesis of 1,2,4-oxadiazoles, 1,3,4-oxadiazoles, and 1,3,4-thiadiazoles from carboxylic acids
  • Activation of the carboxyl group for hydroxyamidation and peptide coupling and in the one-pot conversion of carboxylic acids into hydroxamic acids.t3p is a registered trademark of archimica gmbh

Specifications

CAS 68957-94-8
Density 1.069 g/mL (at 25°C)
Boiling Point 65°C
Molecular Formula C9H21O6P3
Refractive Index n20/D 1.418
Linear Formula C9H21O6P3
MDL Number MFCD00006583
Quantity 500 mL
Synonym 1-Propanephosphonic anhydride solution; 2,4,6-Tripropyl-1,3,5,2,4,6-trioxatriphosphorinane-2,4,6-trioxide solution; PPACA; T3P™
Molecular Weight (g/mol) 318.18
Biological Activity Central Nervous System
Show More Show Less