missing translation for 'onlineSavingsMsg'
Learn More

Propylenediphosphonic acid, 98+%

CAS: 4671-82-3 | C3H10O6P2 | 204.055 g/mol

$56.85 - $538.42

Chemical Identifiers

CAS 4671-82-3
Molecular Formula C3H10O6P2
Molecular Weight (g/mol) 204.055
MDL Number MFCD00050066
InChI Key DZQCAOQMXPROIJ-UHFFFAOYSA-N
Synonym propylenediphosphonic acid, acmc-20ajta, propylene-bisphosphonic acid, propylenediphosphonicacid, propane-1,2-diylbis phosphonic acid, 1-phosphonopropan-2-yl phosphonic acid
PubChem CID 22023450
IUPAC Name 1-phosphonopropan-2-ylphosphonic acid
SMILES CC(CP(=O)(O)O)P(=O)(O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3027702
View Documents
Thermo Scientific Chemicals
03027702
0.5 g
Each for $56.85
Only null left
 
AA3027704
View Documents
Thermo Scientific Chemicals
03027704
2 g
Each for $160.64
Only null left
 
AA3027709
View Documents
Thermo Scientific Chemicals
03027709
10 g
Each for $538.42
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4671-82-3
204.055
DZQCAOQMXPROIJ-UHFFFAOYSA-N
22023450
CC(CP(=O)(O)O)P(=O)(O)O
C3H10O6P2
MFCD00050066
propylenediphosphonic acid, acmc-20ajta, propylene-bisphosphonic acid, propylenediphosphonicacid, propane-1,2-diylbis phosphonic acid, 1-phosphonopropan-2-yl phosphonic acid
1-phosphonopropan-2-ylphosphonic acid

Specifications

4671-82-3
Odorless
MFCD00050066
propylenediphosphonic acid, acmc-20ajta, propylene-bisphosphonic acid, propylenediphosphonicacid, propane-1,2-diylbis phosphonic acid, 1-phosphonopropan-2-yl phosphonic acid
CC(CP(=O)(O)O)P(=O)(O)O
204.055
204.06
Powder
178°C
C3H10O6P2
0.5 g
DZQCAOQMXPROIJ-UHFFFAOYSA-N
1-phosphonopropan-2-ylphosphonic acid
22023450
≥98%
Propylenediphosphonic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only