missing translation for 'onlineSavingsMsg'
Learn More

(±)-Propylene oxide, 99.5%, extra pure

CAS: 75-56-9 | C3H6O | 58.08 g/mol

Supplier:  Thermo Scientific Chemicals 220160025

 View more versions of this product

Catalog No. AC220160025


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(±)-Propylene oxide
75-56-9
0.8300g/mL
−37°C
99.4% min. (GC)
C3H6O
MFCD00005126
1280
0.83
propylene oxide, 1,2-epoxypropane, methyloxirane, epoxypropane, propyleneoxide, propene oxide, oxirane, methyl, 1,2-propylene oxide, propylene epoxide, 2,3-epoxypropane
GOOHAUXETOMSMM-UHFFFAOYNA-N
2-methyloxirane
6378
CHEBI:38685
99.5%
Liquid
Extra Pure
-112.0°C
34.0°C
Authentic
Glass bottle
1.3650 to 1.3670
2.5 L
17, 6
15, 7969
Solubility in water: 40g/100mL (20°C). Other solubilities: miscible with alcohol and ether
CC1CO1
58.08
0.32 mPa.s (20°C)
58.08
Extra Pure

Chemical Identifiers

75-56-9
58.08
GOOHAUXETOMSMM-UHFFFAOYNA-N
6378
2-methyloxirane
C3H6O
MFCD00005126
propylene oxide, 1,2-epoxypropane, methyloxirane, epoxypropane, propyleneoxide, propene oxide, oxirane, methyl, 1,2-propylene oxide, propylene epoxide, 2,3-epoxypropane
CHEBI:38685
CC1CO1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Extremely flammable liquid and vapor.
Harmful if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
May cause cancer.

GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi

GHS Signal Word: Danger

EINECSNumber : 200-879-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only