missing translation for 'onlineSavingsMsg'
Learn More

Propylene Glycol, USP, 99.5%, Spectrum™ Chemical

Catalog No. p-7158495
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605910 500 mL Poly Bottle
18605915 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18605910 Supplier Spectrum Chemical Mfg Cor Supplier No. PR130500MLPL
Only null left

PR130, 57-55-6, C3H8O2

Spectrum™ Chemical Propylene Glycol, USP is used in pharmaceutical products as an excipient solvent in oral and topical drug products. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 57-55-6
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.10
InChI Key DNIAPMSPPWPWGF-UHFFFAOYNA-N
IUPAC Name propane-1,2-diol
SMILES CC(O)CO

Specifications

Acidity 0.20 mL
CAS Max % 1
Ignition Residue 3.5 mg/50g
Packaging Poly Bottle
Quantity 500 mL
Percent Purity 99.5%
Grade USP