missing translation for 'onlineSavingsMsg'
Learn More

Propylene Glycol, U.S.P. - F.C.C., J.T. Baker™

Catalog No. p-5391331
Change view
Click to view available options
Quantity:
500 mL
4 L
19 L
Packaging:
Narrow Mouth Amber Glass Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02003852 500 mL Narrow Mouth Amber Glass Bottle
02003850 4 L Narrow Mouth Amber Glass Bottle
02003848 19 L Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02003852 Supplier Avantor Supplier No. 940201
Only null left

Chemical Identifiers

CAS 57-55-6
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.095
InChI Key DNIAPMSPPWPWGF-UHFFFAOYSA-N
Synonym 1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrost
PubChem CID 1030
ChEBI CHEBI:16997
IUPAC Name propane-1,2-diol
SMILES CC(CO)O

Specifications

Quantity 500 mL
Formula Weight 76.1
Flash Point 99.00°C
Grade FCC/USP
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.04kg
Chemical Name or Material Propylene Glycol