missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Propylene Glycol Monomethyl Ether Acetate
Description
Propylene glycol monomethyl ether acetate (PGMEA) is a photoresist solvent. Its degradation by microorganisms in different soil types has been investigated. An oral reference dose (RfD) value of PGMEA has been obtained from inhalation studies. The solubility of (5-alkylsulfonyloxyimino-5H-thiophen-2-ylidene)-2-methylphenyl-acetonitriles in PGMEA has been analyzed.Propylene glycol monomethyl ether acetate has been used in the development of fluidic channels to be incorporated in a poly(dimethylsiloxane) (PDMS)-based microfluidic device.ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC
Specifications
Specifications
| CAS | 108-65-6 |
| Density | 0.970 g/mL at 25°C |
| Boiling Point | 145°C to 146°C; 145°C to 146°C |
| Molecular Formula | C6H12O3 |
| Linear Formula | CH3CO2CH(CH3)CH2OCH3 |
| Refractive Index | n20/D 1.402 |
| MDL Number | MFCD00038500 |
| Quantity | 4 L |
| Synonym | DOWANOL(R) PMA; 1,2-Propanediol monomethyl ether acetate; 1-Methoxy-2-propyl acetate; MPA; PGMEA; Propylene glycol methyl ether acetate |
| Molecular Weight (g/mol) | 132.16 g/mol |