missing translation for 'onlineSavingsMsg'
Learn More

Propylene Glycol Methyl Ether, 97%, Spectrum™ Chemical

Catalog No. p-7166098
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
55 gal.
Packaging:
Amber Glass Bottle
Drum
Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612976 1 L Amber Glass Bottle
18612977 4 L Amber Glass Bottle
18612974 20 L Steel Can
18612975 55 gal. Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18612976 Supplier Spectrum Chemical Mfg Cor Supplier No. P14381LT
Only null left

Chemical Identifiers

CAS 107-98-2

Specifications

CAS Max % 1
Quantity 1 L
Percent Purity 97%
Grade Reagent
Packaging Amber Glass Bottle
Water 0.001

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.