missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propylene Glycol, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
Supplier: Avantor J.T.Baker U51009
Specifications
| Propylene Glycol | |
| C3H8O2 | |
| 4 L | |
| CC(CO)O | |
| 76.095 | |
| CHEBI:16997 | |
| 99.00°C | |
| Narrow Mouth Amber Glass Bottle |
| 57-55-6 | |
| 1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrost | |
| DNIAPMSPPWPWGF-UHFFFAOYSA-N | |
| propane-1,2-diol | |
| 1030 | |
| 76.1 | |
| BAKER ANALYZED™ ACS Reagent | |
| 1L = 1.04kg |
Chemical Identifiers
| 57-55-6 | |
| 76.095 | |
| 1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrost | |
| CHEBI:16997 | |
| CC(CO)O |
| C3H8O2 | |
| DNIAPMSPPWPWGF-UHFFFAOYSA-N | |
| 1030 | |
| propane-1,2-diol |