missing translation for 'onlineSavingsMsg'
Learn More

Propylene Carbonate, 99%, Spectrum™ Chemical

Catalog No. 18612966
Change view
Click to view available options
Quantity:
25 g
500 g
3 kg
20 kg
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612966 25 g Amber Glass Bottle
18612970 500 g Amber Glass Bottle
18612969 200 L Drum
18612967 3 kg Amber Glass Bottle
18612968 20 kg Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18612966 Supplier Spectrum Chemical Mfg Cor Supplier No. P143325GM

Chemical Identifiers

CAS 108-32-7

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity 0.99
Grade Reagent

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.