missing translation for 'onlineSavingsMsg'
Learn More

Propylamine 98.0+%, TCI America™

Catalog No. p-7134657
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P052025ML 25 mL
P0520500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P052025ML Supplier TCI America Supplier No. P052025ML
Only null left

Chemical Identifiers

CAS 107-10-8
Molecular Formula C3H9N
Molecular Weight (g/mol) 59.112
MDL Number MFCD00008205
InChI Key WGYKZJWCGVVSQN-UHFFFAOYSA-N
Synonym propylamine, 1-propanamine, 1-propylamine, propanamine, n-propylamine, 1-aminopropane, monopropylamine, mono-n-propylamine, propyl amine, n-propyl amine
PubChem CID 7852
ChEBI CHEBI:39870
IUPAC Name propan-1-amine
SMILES CCCN

Specifications

Melting Point -83°C
Color Yellow
pH 12.6 (100g/L H2O 20°C)
Boiling Point 48°C
Quantity 25 mL
UN Number 1277
Formula Weight 59.11
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Propylamine
EINECSNumber (2)-0131
RTECSNumber UH9100000
TSCA Yes