missing translation for 'onlineSavingsMsg'
Learn More

Propylamine Hydrochloride 98.0+%, TCI America™

Catalog No. P052225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P052225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P052225G Supplier TCI America Supplier No. P052225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 556-53-6
Molecular Formula C3H10ClN
Molecular Weight (g/mol) 95.57
MDL Number MFCD00012911
InChI Key PYNUOAIJIQGACY-UHFFFAOYSA-N
Synonym propylamine hydrochloride, n-propylamine hydrochloride, 1-propanamine, hydrochloride, propylammonium chloride, propan-1-amine hydrochloride, 1-propylammonium chloride, n-propylammonium chloride, propylamine, hydrochloride, unii-a1z6ovy2hg, a1z6ovy2hg
PubChem CID 11165
IUPAC Name hydrogen propan-1-amine chloride
SMILES [H+].[Cl-].CCCN

Specifications

Melting Point 164°C
Color White
Quantity 25 g
Formula Weight 95.57
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Propylamine Hydrochloride
EINECSNumber (2)-0131
RTECSNumber UI3057000
TSCA Yes