missing translation for 'onlineSavingsMsg'
Learn More

Propylamine hydrochloride, 98%

Catalog No. AC221890250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC221890250 25 g Glass bottle
AC221891000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC221890250 Supplier Thermo Scientific Chemicals Supplier No. 221890250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 556-53-6 | C3H10ClN | 95.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 556-53-6
Molecular Formula C3H10ClN
Molecular Weight (g/mol) 95.57
MDL Number MFCD00012911
InChI Key PYNUOAIJIQGACY-UHFFFAOYSA-N
Synonym propylamine hydrochloride, n-propylamine hydrochloride, 1-propanamine, hydrochloride, propylammonium chloride, propan-1-amine hydrochloride, 1-propylammonium chloride, n-propylammonium chloride, propylamine, hydrochloride, unii-a1z6ovy2hg, a1z6ovy2hg
PubChem CID 11165
IUPAC Name propan-1-amine;hydrochloride
SMILES [H+].[Cl-].CCCN

Specifications

Melting Point 160°C to 164°C
Color Beige to White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula CH3CH2CH2NH2·HCl
Quantity 25 g
Beilstein 04, 137
Merck Index 15, 7956
Formula Weight 95.57
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Propylamine hydrochloride, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-129-9
RTECSNumber UI3057000
TSCA TSCA

RUO – Research Use Only