missing translation for 'onlineSavingsMsg'
Learn More

Propylamine, 99+%, extra pure

Catalog No. AC220610500
Change view
Click to view available options
Quantity:
50 mL
500 mL
1 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC220610500 50 mL Glass bottle
AC220615000 500 mL Glass bottle
AC220610010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC220610500 Supplier Thermo Scientific Chemicals Supplier No. 220610500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 107-10-8 | C3H9N | 59.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 107-10-8
Molecular Formula C3H9N
Molecular Weight (g/mol) 59.11
MDL Number MFCD00008205
InChI Key WGYKZJWCGVVSQN-UHFFFAOYSA-N
Synonym propylamine, 1-propanamine, 1-propylamine, propanamine, n-propylamine, 1-aminopropane, monopropylamine, mono-n-propylamine, propyl amine, n-propyl amine
PubChem CID 7852
ChEBI CHEBI:39870
IUPAC Name propan-1-amine
SMILES CCCN

Specifications

Melting Point -83.0°C
Density 0.7190g/mL
Boiling Point 48.0°C
Flash Point −30°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.3870 to 1.3890
Linear Formula CH3CH2CH2NH2
Quantity 50 mL
Beilstein 04,136
Fieser 13,164
Merck Index 15,7956
Specific Gravity 0.719
Solubility Information Solubility in water: soluble in water. Other solubilities: miscible with alcohol and ether
Viscosity 0.4 mPa.s (20°C)
Formula Weight 59.11
Percent Purity 99+%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Propylamine, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-462-3
RTECSNumber UH9100000
TSCA TSCA

RUO – Research Use Only