missing translation for 'onlineSavingsMsg'
Learn More

Propyl Propionate 98.0+%, TCI America™

Supplier:  TCI America P0511500ML

 View more versions of this product

Catalog No. P0511500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Propyl Propionate
Colorless
C6H12O2
500 mL
propyl propionate, n-propyl propionate, propanoic acid, propyl ester, n-propyl propanoate, propionic acid, propyl ester, n-propyl n-propionate, propylester kyseliny propionove, unii-g09trv00gk, propyl propionate natural, propionic acid n-propyl ester
CCCOC(=O)CC
116.16
116.16
Liquid
106-36-5
122°C
MFCD00009373
3272
MCSINKKTEDDPNK-UHFFFAOYSA-N
propyl propanoate
7803
≥98.0% (GC)

Chemical Identifiers

106-36-5
116.16
MCSINKKTEDDPNK-UHFFFAOYSA-N
7803
CCCOC(=O)CC
C6H12O2
MFCD00009373
propyl propionate, n-propyl propionate, propanoic acid, propyl ester, n-propyl propanoate, propionic acid, propyl ester, n-propyl n-propionate, propylester kyseliny propionove, unii-g09trv00gk, propyl propionate natural, propionic acid n-propyl ester
propyl propanoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0774

RTECSNumber : UF7100000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions