missing translation for 'onlineSavingsMsg'
Learn More

Propyl Propionate 98.0+%, TCI America™

Catalog No. P051125ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P051125ML 25 mL
P0511500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P051125ML Supplier TCI America Supplier No. P051125ML

Chemical Identifiers

CAS 106-36-5
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009373
InChI Key MCSINKKTEDDPNK-UHFFFAOYSA-N
Synonym propyl propionate, n-propyl propionate, propanoic acid, propyl ester, n-propyl propanoate, propionic acid, propyl ester, n-propyl n-propionate, propylester kyseliny propionove, unii-g09trv00gk, propyl propionate natural, propionic acid n-propyl ester
PubChem CID 7803
IUPAC Name propyl propanoate
SMILES CCCOC(=O)CC

Specifications

Color Colorless
Boiling Point 122°C
Quantity 25 mL
UN Number 3272
Formula Weight 116.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Propyl Propionate
EINECSNumber (2)-0774
RTECSNumber UF7100000
TSCA Yes