missing translation for 'onlineSavingsMsg'
Learn More

Propyl p-Toluenesulfonate 98.0+%, TCI America™

Catalog No. p-7136410
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
T072625ML 25 mL
T0726500ML 500 mL
2 options

Catalog No. T072625ML

Supplier: TCI America T072625ML

Only null left

Chemical Identifiers

CAS 599-91-7
Molecular Formula C10H14O3S
Molecular Weight (g/mol) 214.279
MDL Number MFCD00059244
InChI Key JTTWNTXHFYNETH-UHFFFAOYSA-N
Synonym propyl p-toluenesulfonate, propyl tosylate, benzenesulfonic acid, 4-methyl-, propyl ester, p-toluenesulfonic acid, propyl ester, propyl toluene-4-sulphonate, ccris 9150, p-toluenesulfonic acid n-propyl ester, p-toluenesulfonic acid propyl ester, propanol tosylate, acmc-209mha
PubChem CID 69035
IUPAC Name propyl 4-methylbenzenesulfonate
SMILES CCCOS(=O)(=O)C1=CC=C(C=C1)C

Specifications

Color Yellow
Boiling Point 156°C
Quantity 25 mL
Formula Weight 214.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Propyl p-Toluenesulfonate
EINECSNumber (3)-1896
TSCA Yes