missing translation for 'onlineSavingsMsg'
Learn More

Propyl p-Toluenesulfonate 98.0+%, TCI America™

Catalog No. T072625ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T072625ML 25 mL
T0726500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T072625ML Supplier TCI America Supplier No. T072625ML

Chemical Identifiers

CAS 599-91-7
Molecular Formula C10H14O3S
Molecular Weight (g/mol) 214.279
MDL Number MFCD00059244
InChI Key JTTWNTXHFYNETH-UHFFFAOYSA-N
Synonym propyl p-toluenesulfonate, propyl tosylate, benzenesulfonic acid, 4-methyl-, propyl ester, p-toluenesulfonic acid, propyl ester, propyl toluene-4-sulphonate, ccris 9150, p-toluenesulfonic acid n-propyl ester, p-toluenesulfonic acid propyl ester, propanol tosylate, acmc-209mha
PubChem CID 69035
IUPAC Name propyl 4-methylbenzenesulfonate
SMILES CCCOS(=O)(=O)C1=CC=C(C=C1)C

Specifications

Color Yellow
Boiling Point 156°C
Quantity 25 mL
Formula Weight 214.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Propyl p-Toluenesulfonate
EINECSNumber (3)-1896
TSCA Yes