missing translation for 'onlineSavingsMsg'
Learn More

Propyl Hexanoate 98.0+%, TCI America™

Catalog No. p-7135324
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P194025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P194025ML Supplier TCI America Supplier No. P194025ML
Only null left

Chemical Identifiers

CAS 626-77-7
Molecular Formula C9H18O2
Molecular Weight (g/mol) 158.24
MDL Number MFCD00053803
InChI Key HTUIWRWYYVBCFT-UHFFFAOYSA-N
Synonym propyl caproate, caproic acid propyl ester, hexanoic acid, propyl ester, n-propyl hexanoate, propionyl hexanoate, n-propyl n-hexanoate, hexanoic acid propyl ester, natural, fema no. 2949, n-propyl hexanoaten
PubChem CID 12293
ChEBI CHEBI:87365
IUPAC Name propyl hexanoate
SMILES CCCCCC(=O)OCCC

Specifications

Melting Point -69°C
Color Colorless
Boiling Point 187°C
Quantity 25 mL
UN Number 3272
Formula Weight 158.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Propyl Hexanoate
EINECSNumber (2)-0782
TSCA Yes