missing translation for 'onlineSavingsMsg'
Learn More

Propyl Gallate 98.0+%, TCI America™

Catalog No. G001825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G001825G 25 g
G0018500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G001825G Supplier TCI America Supplier No. G001825G

Chemical Identifiers

CAS 121-79-9
Molecular Formula C10H12O5
Molecular Weight (g/mol) 212.201
MDL Number MFCD00002196
InChI Key ZTHYODDOHIVTJV-UHFFFAOYSA-N
Synonym propyl gallate, n-propyl gallate, progallin p, tenox pg, nipagallin p, gallic acid, propyl ester, gallic acid propyl ester, nipa 49, benzoic acid, 3,4,5-trihydroxy-, propyl ester, 3,4,5-trihydroxybenzene-1-propylcarboxylate
PubChem CID 4947
ChEBI CHEBI:10607
IUPAC Name propyl 3,4,5-trihydroxybenzoate
SMILES CCCOC(=O)C1=CC(=C(C(=C1)O)O)O

Specifications

Melting Point 147°C
Quantity 25 g
Formula Weight 212.2
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Propyl Gallate
RTECSNumber LW8400000
TSCA Yes