missing translation for 'onlineSavingsMsg'
Learn More

Propyl Gallate, NF, 98-102%, Spectrum™ Chemical

Catalog No. 18605902
Change view
Click to view available options
Quantity:
2.5 kg
25 kg
Packaging:
Fiber Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605902 2.5 kg -
18605903 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18605902 Supplier Spectrum Chemical Mfg Cor Supplier No. P14532.5KG
Get Up to $500 in Chemicals with Purchase
View all promotions

P1453, 121-79-9, C10H12O5

Spectrum™ Chemical Propyl Gallate, NF is used as an antioxidant in pharmaceutical formulations. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 121-79-9
Molecular Weight (g/mol) 212.2
InChI Key ZTHYODDOHIVTJV-UHFFFAOYSA-N
IUPAC Name propyl 3,4,5-trihydroxybenzoate
SMILES CCCOC(=O)C1=CC(O)=C(O)C(O)=C1

Specifications

Melting Range 146-150°C
Quantity 2.5 kg
Percent Purity 98 to 102%
Grade NF