missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propyl Ether 98.0+%, TCI America™
$175.22 - $1295.52
Chemical Identifiers
| CAS | 111-43-3 |
|---|---|
| Molecular Formula | C6H14O |
| Molecular Weight (g/mol) | 102.18 |
| MDL Number | MFCD00009376 |
| InChI Key | POLCUAVZOMRGSN-UHFFFAOYSA-N |
| Synonym | dipropyl ether, propyl ether, n-propyl ether, di-n-propyl ether, propane, 1,1'-oxybis, dipropyl oxide, 4-oxaheptane, ether, di-n-propyl, 1,1'-oxybispropane, unii-42q250hs4g |
| PubChem CID | 8114 |
| IUPAC Name | 1-propoxypropane |
| SMILES | CCCOCCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
P052725ML
|
TCI America
P052725ML |
25 mL |
Each for $175.22
|
|
|||||
|
P0527500ML
|
TCI America
P0527500ML |
500 mL |
Each for $1,295.52
|
|
|||||
Chemical Identifiers
| 111-43-3 | |
| 102.18 | |
| POLCUAVZOMRGSN-UHFFFAOYSA-N | |
| 8114 | |
| CCCOCCC |
| C6H14O | |
| MFCD00009376 | |
| dipropyl ether, propyl ether, n-propyl ether, di-n-propyl ether, propane, 1,1'-oxybis, dipropyl oxide, 4-oxaheptane, ether, di-n-propyl, 1,1'-oxybispropane, unii-42q250hs4g | |
| 1-propoxypropane |
Specifications
| 111-43-3 | |
| Colorless | |
| C6H14O | |
| 25 mL | |
| dipropyl ether, propyl ether, n-propyl ether, di-n-propyl ether, propane, 1,1'-oxybis, dipropyl oxide, 4-oxaheptane, ether, di-n-propyl, 1,1'-oxybispropane, unii-42q250hs4g | |
| CCCOCCC | |
| 102.18 | |
| 102.18 | |
| Liquid |
| -123°C | |
| 90°C | |
| MFCD00009376 | |
| 2384 | |
| POLCUAVZOMRGSN-UHFFFAOYSA-N | |
| 1-propoxypropane | |
| 8114 | |
| ≥98.0% (GC) | |
| Propyl Ether |
Safety and Handling
EINECSNumber : (2)-0362
RTECSNumber : UJ5125000
TSCA : Yes