missing translation for 'onlineSavingsMsg'
Learn More

Propyl Ether 98.0+%, TCI America™

Catalog No. P052725ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P052725ML 25 mL
P0527500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P052725ML Supplier TCI America Supplier No. P052725ML

Chemical Identifiers

CAS 111-43-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.18
MDL Number MFCD00009376
InChI Key POLCUAVZOMRGSN-UHFFFAOYSA-N
Synonym dipropyl ether, propyl ether, n-propyl ether, di-n-propyl ether, propane, 1,1'-oxybis, dipropyl oxide, 4-oxaheptane, ether, di-n-propyl, 1,1'-oxybispropane, unii-42q250hs4g
PubChem CID 8114
IUPAC Name 1-propoxypropane
SMILES CCCOCCC

Specifications

Melting Point -123°C
Color Colorless
Boiling Point 90°C
Quantity 25 mL
UN Number 2384
Formula Weight 102.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Propyl Ether
EINECSNumber (2)-0362
RTECSNumber UJ5125000
TSCA Yes