missing translation for 'onlineSavingsMsg'
Learn More

Propyl Butyrate 98.0+%, TCI America™

Catalog No. B076425ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B076425ML 25 mL
B0764500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B076425ML Supplier TCI America Supplier No. B076425ML

Chemical Identifiers

CAS 105-66-8
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
MDL Number MFCD00009396
InChI Key HUAZGNHGCJGYNP-UHFFFAOYSA-N
Synonym propyl butyrate, butanoic acid, propyl ester, propyl n-butyrate, butyric acid, propyl ester, 1-propyl butyrate, n-propyl butyrate, n-propyl n-butyrate, n-propyl-n-butanoate, n-butyric acid n-propyl ester, butyric acid propyl ester
PubChem CID 7770
IUPAC Name propyl butanoate
SMILES CCCC(=O)OCCC

Specifications

Color Colorless
Boiling Point 144°C
Quantity 25 mL
UN Number 3272
Formula Weight 130.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Propyl Butyrate
EINECSNumber (2)-0779
RTECSNumber ET6200000
TSCA Yes