missing translation for 'onlineSavingsMsg'
Learn More

Propyl 3-Propoxypyridine-2-carboxylate 98.0+%, TCI America™

Catalog No. P11965G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11965G 5 g
P11961G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P11965G Supplier TCI America Supplier No. P11965G
Only null left

Chemical Identifiers

CAS 134319-22-5
Molecular Formula C12H17NO3
Molecular Weight (g/mol) 223.272
MDL Number MFCD00191575
InChI Key ZGUCGLNLZXLQTD-UHFFFAOYSA-N
Synonym 3-Propoxypicolinic Acid Propyl Ester, Propyl 3-Propoxypicolinate, 3-Propoxypyridine-2-carboxylic Acid Propyl Ester
PubChem CID 19756120
IUPAC Name propyl 3-propoxypyridine-2-carboxylate
SMILES CCCOC1=C(N=CC=C1)C(=O)OCCC

Specifications

Color Yellow
Quantity 5 g
Formula Weight 223.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Propyl 3-Propoxypyridine-2-carboxylate