missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Propranolol hydrochloride, 99%

Catalog No. AAH2664514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2664514 25 g
AAH2664506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2664514 Supplier Thermo Scientific Chemicals Supplier No. H2664514

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 318-98-9 | C16H22ClNO2 | 295.807 g/mol

A beta-adrenergic blocker

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 318-98-9
Molecular Formula C16H22ClNO2
Molecular Weight (g/mol) 295.807
MDL Number MFCD00012558
InChI Key ZMRUPTIKESYGQW-UHFFFAOYSA-N
Synonym propranolol hydrochloride, propranolol hcl, inderal, avlocardyl, herzbase, inderalici, naprilin, pronovan, dociton, ikopal
PubChem CID 62882
ChEBI CHEBI:8500
IUPAC Name 1-naphthalen-1-yloxy-3-(propan-2-ylamino)propan-2-ol;hydrochloride
SMILES CC(C)NCC(COC1=CC=CC2=CC=CC=C21)O.Cl

Specifications

Melting Point 163°C to 165°C
Quantity 25 g
Merck Index 14,7840
Formula Weight 295.8
Percent Purity 99%
Chemical Name or Material (+/-)-Propranolol hydrochloride
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 206-268-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only