missing translation for 'onlineSavingsMsg'
Learn More

Propofol Impurity J, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111022840
Catalog No. 111022840 Supplier Merck Emd Millipore Supplier No. BP1032

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

Specifications

Flash Point Not Applicable
Molecular Formula C12H16O2
Linear Formula C12H16O2
Synonym 2,6-Diisopropyl-1,4-benzoquinone