missing translation for 'onlineSavingsMsg'
Learn More

Propiophenone 98.0+%, TCI America™

Catalog No. P0519500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0519500G 500 g
P051925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0519500G Supplier TCI America Supplier No. P0519500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 93-55-0
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00009309
InChI Key KRIOVPPHQSLHCZ-UHFFFAOYSA-N
Synonym propiophenone, ethyl phenyl ketone, propionylbenzene, 1-phenyl-1-propanone, 1-propanone, 1-phenyl, phenyl ethyl ketone, ketone, ethyl phenyl, propionphenone, usaf ek-1235, 1-phenyl-propan-1-one
PubChem CID 7148
ChEBI CHEBI:425902
IUPAC Name 1-phenylpropan-1-one
SMILES CCC(=O)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 216°C
Quantity 500 g
Formula Weight 134.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Propiophenone
EINECSNumber (9)-1157&(3)-2673
RTECSNumber UG7175000
TSCA Yes