missing translation for 'onlineSavingsMsg'
Learn More

Propiophenone, 99%

Catalog No. AAA1514022
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1514022 100 g
AAA1514036 500 g
AAA151400E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1514022 Supplier Thermo Scientific Chemicals Supplier No. A1514022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 93-55-0 | C9H10O | 134.178 g/mol

Propiophenone is an intermediate in the synthesis of pharmaceuticals and organic compounds. It is used in the synthesis of ephedrine and propiophenone derivatives such as cathinone, and methcathinone. It can also be used in the synthesis of aryl alkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propiophenone is an intermediate in the synthesis of pharmaceuticals and organic compounds. It is used in the synthesis of ephedrine and propiophenone derivatives such as cathinone, and methcathinone. It can also be used in the synthesis of aryl alkenes.

Solubility
Difficult to mix.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 93-55-0
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00009309
InChI Key KRIOVPPHQSLHCZ-UHFFFAOYSA-N
Synonym propiophenone, ethyl phenyl ketone, propionylbenzene, 1-phenyl-1-propanone, 1-propanone, 1-phenyl, phenyl ethyl ketone, ketone, ethyl phenyl, propionphenone, usaf ek-1235, 1-phenyl-propan-1-one
PubChem CID 7148
ChEBI CHEBI:425902
IUPAC Name 1-phenylpropan-1-one
SMILES CCC(=O)C1=CC=CC=C1

Specifications

Melting Point 17°C to 18°C
Density 1.008
Boiling Point 217°C to 218°C
Flash Point 87°C (188°F)
Refractive Index 1.526
Quantity 100 g
Beilstein 606215
Merck Index 14,7830
Solubility Information Difficult to mix.
Formula Weight 134.18
Percent Purity 99%
Chemical Name or Material Propiophenone
Show More Show Less
Hazard Category H227-H319
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P264b-P280-P305+P351+P338-P370+P378q-P501c
EINECSNumber 202-257-6
RTECSNumber UG7175000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only