missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propionyl-L-carnitine Hydrochloride 98.0+%, TCI America™
$72.70 - $734.44
Chemical Identifiers
| CAS | 119793-66-7 |
|---|---|
| Molecular Formula | C10H20ClNO4 |
| Molecular Weight (g/mol) | 253.72 |
| MDL Number | MFCD00274137 |
| InChI Key | KTFMPDDJYRFWQE-DDWIOCJRSA-N |
| Synonym | (R)-3-Propionyloxy-4-(trimethylammonio)butyrate Hydrochloride |
| PubChem CID | 129893533 |
| IUPAC Name | [(2R)-3-carboxy-2-(propanoyloxy)propyl]trimethylazanium chloride |
| SMILES | [Cl-].CCC(=O)O[C@H](CC(O)=O)C[N+](C)(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
P2569100MG
|
TCI America
P2569100MG |
100 mg |
Each for $72.70
|
|
|||||
|
P25691G
|
TCI America
P25691G |
1 g |
Each for $734.44
|
|
|||||
Chemical Identifiers
| 119793-66-7 | |
| 253.72 | |
| KTFMPDDJYRFWQE-DDWIOCJRSA-N | |
| 129893533 | |
| [Cl-].CCC(=O)O[C@H](CC(O)=O)C[N+](C)(C)C |
| C10H20ClNO4 | |
| MFCD00274137 | |
| (R)-3-Propionyloxy-4-(trimethylammonio)butyrate Hydrochloride | |
| [(2R)-3-carboxy-2-(propanoyloxy)propyl]trimethylazanium chloride |
Specifications
| 119793-66-7 | |
| White | |
| MFCD00274137 | |
| (R)-3-Propionyloxy-4-(trimethylammonio)butyrate Hydrochloride | |
| [Cl-].CCC(=O)O[C@H](CC(O)=O)C[N+](C)(C)C | |
| 253.72 | |
| 253.72 | |
| Crystalline Powder |
| 168°C | |
| C10H20ClNO4 | |
| 100 mg | |
| KTFMPDDJYRFWQE-DDWIOCJRSA-N | |
| [(2R)-3-carboxy-2-(propanoyloxy)propyl]trimethylazanium chloride | |
| 129893533 | |
| ≥98.0% (T) | |
| Propionyl-L-carnitine Hydrochloride |
Safety and Handling
TSCA : No
Recommended Storage : Freezer (-20 °C)