missing translation for 'onlineSavingsMsg'
Learn More

Propionyl-L-carnitine Hydrochloride 98.0+%, TCI America™

Catalog No. P2569100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2569100MG 100 mg
P25691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2569100MG Supplier TCI America Supplier No. P2569100MG

Chemical Identifiers

CAS 119793-66-7
Molecular Formula C10H20ClNO4
Molecular Weight (g/mol) 253.72
MDL Number MFCD00274137
InChI Key KTFMPDDJYRFWQE-DDWIOCJRSA-N
Synonym (R)-3-Propionyloxy-4-(trimethylammonio)butyrate Hydrochloride
PubChem CID 129893533
IUPAC Name [(2R)-3-carboxy-2-(propanoyloxy)propyl]trimethylazanium chloride
SMILES [Cl-].CCC(=O)O[C@H](CC(O)=O)C[N+](C)(C)C

Specifications

Melting Point 168°C
Color White
Quantity 100 mg
Formula Weight 253.72
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Propionyl-L-carnitine Hydrochloride
TSCA No
Recommended Storage Freezer (-20 °C)