missing translation for 'onlineSavingsMsg'
Learn More

Propionyl Bromide 98.0+%, TCI America™

Catalog No. P051525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P051525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P051525G Supplier TCI America Supplier No. P051525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 598-22-1
Molecular Formula C3H5BrO
Molecular Weight (g/mol) 136.976
MDL Number MFCD00000116
InChI Key RIBFXMJCUYXJDZ-UHFFFAOYSA-N
Synonym propionyl bromide, propanoyl-bromide, propionylbromide, propanyl bromide, acmc-209mg3, ksc270q9p, ribfxmjcuyxjdz-uhfffaoysa, inchi=1/c3h5bro/c1-2-3 4 5/h2h2,1h3
PubChem CID 69018
IUPAC Name propanoyl bromide
SMILES CCC(=O)Br

Specifications

Color Yellow
Boiling Point 104°C
Quantity 25 g
UN Number 2920
Formula Weight 136.98
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Propionyl Bromide