missing translation for 'onlineSavingsMsg'
Learn More

Propionic Acid 99.0+%, TCI America™

Catalog No. P0500500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0500500ML 500 mL
P050025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0500500ML Supplier TCI America Supplier No. P0500500ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 79-09-4
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.079
MDL Number MFCD00002756
InChI Key XBDQKXXYIPTUBI-UHFFFAOYSA-N
Synonym propionic acid, ethylformic acid, methylacetic acid, carboxyethane, ethanecarboxylic acid, pseudoacetic acid, metacetonic acid, monoprop, luprosil, prozoin
PubChem CID 1032
ChEBI CHEBI:30768
IUPAC Name propanoic acid
SMILES CCC(=O)O

Specifications

Color Colorless
Boiling Point 140°C
Quantity 500 mL
UN Number 3463
Formula Weight 74.08
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Propionic Acid
EINECSNumber (2)-0602
RTECSNumber UE5950000
TSCA Yes