missing translation for 'onlineSavingsMsg'
Learn More

Propionic Acid, NF, 99.5-100.5%, Spectrum™ Chemical

Catalog No. 18605887
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605887 200 L Drum
18605883 500 mL Amber Glass Bottle
18605882 1 L Amber Glass Bottle
18605884 4 L Amber Glass Bottle
18605886 20 L Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18605887 Supplier Spectrum Chemical Mfg Cor Supplier No. P1464200LTBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

P1464, 79-09-4, C3H6O2

Spectrum™ Chemical Propionic Acid, NF is used as a preservative in pharmaceutical formulations. Spectrum Chemical manufactured NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

CAS 79-09-4
Packaging Drum
Quantity 200 L
Percent Purity 99.5 to 100.5%