missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propionic Acid, FCC, 99.5-100.5%, Spectrum™ Chemical
Description
Spectrum™ Chemical Propionic Acid, FCC is used as a preservative and flavoring agent in packaged foods such as baked goods and cheese. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 1 |
| Packaging | Drum |
| Quantity | 200 L |
| Specific Gravity | 0.993-0.997 |
| Percent Purity | 99.5 to 100.5% |
| Grade | FCC |
| Physical Form | Liquid |