missing translation for 'onlineSavingsMsg'
Learn More

Propionic Acid (Certified ACS), Fisher Chemical

Catalog No. A258500
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A258500 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A258500 Supplier Thermo Fisher Scientific Supplier No. BPA258500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 79-09-4
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.079
MDL Number MFCD00002756
InChI Key XBDQKXXYIPTUBI-UHFFFAOYSA-N
Synonym propionic acid, ethylformic acid, methylacetic acid, carboxyethane, ethanecarboxylic acid, pseudoacetic acid, metacetonic acid, monoprop, luprosil, prozoin
PubChem CID 1032
ChEBI CHEBI:30768
IUPAC Name propanoic acid
SMILES CCC(=O)O

Specifications

Boiling Range 3°C max. (1 to 95mL)
Melting Point -22°C
Density 0.990 g/cm3
pH 2.5
Boiling Point 141°C
Flash Point 52°C
Packaging Glass Bottle
Linear Formula CH3CH2COOH
Quantity 500 mL
UN Number 1848
Residue after Evaporation 0.01% max.
Vapor Density 2.56g/mol
Vapor Pressure 5mbar at 20°C
Formula Weight 74.08
Grade Certified
Physical Form Liquid
Chemical Name or Material Propionic Acid
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
DOTInformation Class 8: Corrosive