missing translation for 'onlineSavingsMsg'
Learn More

Propionic acid, 99%

Catalog No. p-7044043
Change view
Click to view available options
Quantity:
100 g
1000 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0421022 100 g
AAL042100B 1000 g
AAL042100E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0421022 Supplier Thermo Scientific Chemicals Supplier No. L0421022
Only null left

CAS: 79-09-4 | C3H6O2 | 74.079 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Propionic acid is used as a preservative for animal feed and food for human consumption. It acts as an intermediate in the production of polymers like cellulose-acetate-propionate, artificial flavorings, agrochemicals and in bleaching agents. In addition, it is used in the synthesis of morphans.

Solubility
Miscible with water, alcohol, diethyl ether and chloroform.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 79-09-4
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.079
MDL Number MFCD00002756
InChI Key XBDQKXXYIPTUBI-UHFFFAOYSA-N
Synonym propionic acid, ethylformic acid, methylacetic acid, carboxyethane, ethanecarboxylic acid, pseudoacetic acid, metacetonic acid, monoprop, luprosil, prozoin
PubChem CID 1032
ChEBI CHEBI:30768
IUPAC Name propanoic acid
SMILES CCC(=O)O

Specifications

Melting Point -21°C
Density 0.992
Boiling Point 140°C to 141°C
Flash Point 51°C (123°F)
Odor Unpleasant
Refractive Index 1.3875
Linear Formula CH3CH2CO2H
Quantity 100 g
UN Number UN3463
Beilstein 506071
Merck Index 14,7825
Solubility Information Miscible with water,alcohol,diethyl ether and chloroform.
Formula Weight 74.08
Percent Purity 99%
Chemical Name or Material Propionic acid
Show More Show Less
Hazard Category H226-H290-H314-H335
Hazard Statement GHS H Statement
H314-H226
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P234-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: PROPIONIC ACID
EINECSNumber 201-176-3
RTECSNumber UE5950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only