missing translation for 'onlineSavingsMsg'
Learn More

Propionic acid, 99+%, extra pure

Catalog No. AC220130010
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC220130010 1 L Glass bottle
AC220130025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC220130010 Supplier Thermo Scientific Chemicals Supplier No. 220130010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 79-09-4 | C3H6O2 | 74.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79-09-4
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.08
MDL Number MFCD00002756
InChI Key XBDQKXXYIPTUBI-UHFFFAOYSA-N
Synonym propionic acid, ethylformic acid, methylacetic acid, carboxyethane, ethanecarboxylic acid, pseudoacetic acid, metacetonic acid, monoprop, luprosil, prozoin
PubChem CID 1032
ChEBI CHEBI:30768
IUPAC Name propanoic acid
SMILES CCC(=O)O

Specifications

Melting Point -22.0°C
Density 0.9900g/mL
Boiling Point 141.0°C
Flash Point 51°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.3850 to 1.3870
Linear Formula C2H5CO2H
Quantity 1 L
Beilstein 02, 234
Fieser 12,415
Specific Gravity 0.99
Merck Index 15, 7939
Solubility Information Solubility in water: soluble.
Viscosity 1.02 mPa.s (25°C)
Formula Weight 74.08
Percent Purity 99+%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Propionic acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapour.
May be corrosive to metals.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
EINECSNumber 201-176-3

RUO – Research Use Only