missing translation for 'onlineSavingsMsg'
Learn More

Propionamide 98.0+%, TCI America™

Catalog No. P049925G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P049925G 25 g
P0499500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P049925G Supplier TCI America Supplier No. P049925G

Chemical Identifiers

CAS 79-05-0
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.095
MDL Number MFCD00008039
InChI Key QLNJFJADRCOGBJ-UHFFFAOYSA-N
Synonym propionamide, propylamide, propionic amide, propionimidic acid, propanimidic acid, propionic acid amide, amid kyseliny propionove, propanimidic acid van, propionimidic acid van, unii-qk07g0hp47
PubChem CID 6578
ChEBI CHEBI:45422
IUPAC Name propanamide
SMILES CCC(=O)N

Specifications

Melting Point 78°C
Color White
Boiling Point 213°C
Quantity 25 g
Formula Weight 73.10
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material Propionamide
RTECSNumber UE2975000
TSCA Yes