missing translation for 'onlineSavingsMsg'
Learn More

Propionaldehyde 98.0+%, TCI America™

Catalog No. P0498500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0498500ML 500 mL
P049825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0498500ML Supplier TCI America Supplier No. P0498500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 123-38-6
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00007020
InChI Key NBBJYMSMWIIQGU-UHFFFAOYSA-N
Synonym propionaldehyde, propanaldehyde, propaldehyde, propional, propionic aldehyde, n-propanal, methylacetaldehyde, propylaldehyde, propylic aldehyde, aldehyde propionique
PubChem CID 527
ChEBI CHEBI:17153
IUPAC Name propanal
SMILES CCC=O

Specifications

Melting Point -81°C
Color Colorless
Boiling Point 46°C
Quantity 500 mL
UN Number 1275
Formula Weight 58.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Propionaldehyde
EINECSNumber (2)-0486
RTECSNumber UE0350000
TSCA Yes
Recommended Storage Refrigerator