Learn More
Propionaldehyde, 97%
Description
Propionaldehyde applications include as an intermediate, in pharmaceuticals, perfumes, plastics and polyols or polyhydric alcohols. It is also used as a precursor to trimethylolethane (CH3C(CH2OH)3) through a condensation reaction with formaldehyde, this triol is an important intermediate in the production of alkyd resins. Other applications include reduction to propanol and oxidation to propionic acid. Also used in PEGylated protein purification.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Propionaldehyde applications include as an intermediate, in pharmaceuticals, perfumes, plastics and polyols or polyhydric alcohols. It is also used as a precursor to trimethylolethane (CH3C(CH2OH)3) through a condensation reaction with formaldehyde, this triol is an important intermediate in the production of alkyd resins. Other applications include reduction to propanol and oxidation to propionic acid. Also used in PEGylated protein purification.
Solubility
Fully miscible in water.
Notes
Air sensitive. Incompatible with oxidizing agents and heat.
Specifications
Specifications
| Melting Point | -81°C |
| Density | 0.81 |
| Boiling Point | 47°C to 49°C |
| Flash Point | −40°C (−40°F) |
| Odor | Pungent |
| Refractive Index | 1.362 |
| Quantity | 500 mL |
| UN Number | UN1275 |
| Beilstein | 506010 |
| Sensitivity | Air sensitive |
| Show More |
Safety and Handling
| Hazard Category | H225-H302+H332-H315-H318-H335 |
| Hazard Statement | GHS H Statement H225-H315-H319-H335 Highly flammable liquid and vapour. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PROPIONALDEHYDE |
| EINECSNumber | 204-623-0 |
| RTECSNumber | UE0350000 |
| TSCA | Yes |
| Recommended Storage | Keep cold; Store under Argon |
RUO – Research Use Only