missing translation for 'onlineSavingsMsg'
Learn More

Propionaldehyde, 97%, Spectrum™ Chemical

Catalog No. p-7166095
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612957 100 mL Amber Glass Bottle
18612959 500 mL Amber Glass Bottle
18612958 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18612957 Supplier Spectrum Chemical Mfg Cor Supplier No. P1432100ML
Only null left

Chemical Identifiers

CAS 123-38-6

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 100 mL
Percent Purity 0.97
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.