Learn More
Propiolamide, 96%
CAS: 7341-96-0 | C3H3NO | 69.06 g/mol
Supplier: Thermo Scientific Chemicals H5231906
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Propiolamide | |
| C3H3NO | |
| MFCD04035573 | |
| propiolamide, 2-propynamide, propynoic acid amide, propynamide, propiolic acid amide, propargylamide, hc.equiv.cconh2, propynoicacidamide, 2-propynamide 9ci | |
| NC(=O)C#C | |
| 69.06 | |
| CHEBI:51764 | |
| 96% |
| 7341-96-0 | |
| HC≡CCONH2 | |
| 5 g | |
| HCJTYESURSHXNB-UHFFFAOYSA-N | |
| prop-2-ynamide | |
| 101445 | |
| 69.06 |
Chemical Identifiers
| 7341-96-0 | |
| 69.06 | |
| HCJTYESURSHXNB-UHFFFAOYSA-N | |
| 101445 | |
| prop-2-ynamide |
| C3H3NO | |
| MFCD04035573 | |
| propiolamide, 2-propynamide, propynoic acid amide, propynamide, propiolic acid amide, propargylamide, hc.equiv.cconh2, propynoicacidamide, 2-propynamide 9ci | |
| CHEBI:51764 | |
| NC(=O)C#C |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only