missing translation for 'onlineSavingsMsg'
Learn More

Propiolaldehyde diethyl acetal, 97%

Catalog No. p-7045448
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1076703 1 g
AAL1076706 5 g
AAL1076714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1076703 Supplier Thermo Scientific Chemicals Supplier No. L1076703

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10160-87-9 | C7H12O2 | 128.17 g/mol

Propiolaldehyde diethyl acetal is used in preparation of 3-boronoacrolein pinacolate, a heterodyne and (E)-3-(tributylstannyl)-2-propenal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propiolaldehyde diethyl acetal is used in preparation of 3-boronoacrolein pinacolate, a heterodyne and (E)-3-(tributylstannyl)-2-propenal.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. Air and moisture sensitive. Store under inert gas.

Chemical Identifiers

CAS 10160-87-9
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00009237
InChI Key RGUXEWWHSQGVRZ-UHFFFAOYSA-N
PubChem CID 66285
IUPAC Name 3,3-diethoxyprop-1-yne
SMILES CCOC(OCC)C#C

Specifications

Density 0.894
Boiling Point 138°C to 139°C
Flash Point 32°C (89°F)
Refractive Index 1.412
Quantity 1 g
UN Number UN1993
Beilstein 1701566
Synonym 3,3-diethoxy-1-propyne, propargylaldehyde diethyl acetal, 3,3-diethoxypropyne, propiolaldehyde diethyl acetal, 1-propyne, 3,3-diethoxy, 3,3-diethoxy-propyne, propynal diethyl acetal, 1,1-diethoxyprop-2-yne, 1-propyne,3,3-diethoxy, propargyl aldehyde diethylacetal
Solubility Information Slightly soluble in water.
Formula Weight 128.17
Percent Purity 97%
Chemical Name or Material Propiolaldehyde diethyl acetal
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 233-430-4
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only