Learn More
Propargyltrimethylsilane, 80-90%, stabilized
CAS: 13361-64-3 | C6H12Si | 112.25 g/mol
$431.92 - $2359.28
Chemical Identifiers
| CAS | 13361-64-3 |
|---|---|
| Molecular Formula | C6H12Si |
| Molecular Weight (g/mol) | 112.25 |
| MDL Number | MFCD00042922 |
| InChI Key | ULYLMHUHFUQKOE-UHFFFAOYSA-N |
| Synonym | propargyltrimethylsilane, silane, trimethyl-2-propynyl, trimethyl 2-propynyl silane, trimethyl prop-2-ynyl silane, trimethyl propargyl silane, silane, trimethyl-2-propyn-1-yl, trimethyl prop-2-yn-1-yl silane, trimethyl-2-propynylsilane, propargyl trimethylsilane, 2-propynyltrimethylsilane |
| PubChem CID | 83378 |
| IUPAC Name | trimethyl(prop-2-ynyl)silane |
| SMILES | C[Si](C)(C)CC#C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC366630010
|
Thermo Scientific Chemicals
366630010 |
1 g | Glass bottle |
Each for $431.92
|
|
||||
|
AC366630050
|
Thermo Scientific Chemicals
366630050 |
5 g | Glass bottle |
Each for $2,359.28
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 13361-64-3 | |
| 112.25 | |
| ULYLMHUHFUQKOE-UHFFFAOYSA-N | |
| 83378 | |
| C[Si](C)(C)CC#C |
| C6H12Si | |
| MFCD00042922 | |
| propargyltrimethylsilane, silane, trimethyl-2-propynyl, trimethyl 2-propynyl silane, trimethyl prop-2-ynyl silane, trimethyl propargyl silane, silane, trimethyl-2-propyn-1-yl, trimethyl prop-2-yn-1-yl silane, trimethyl-2-propynylsilane, propargyl trimethylsilane, 2-propynyltrimethylsilane | |
| trimethyl(prop-2-ynyl)silane |
Specifications
| 13361-64-3 | |
| 0.7500g/mL | |
| -10°C | |
| 80 to 90% (GC) | |
| C6H12Si | |
| MFCD00042922 | |
| 04, IV, 3938 | |
| propargyltrimethylsilane, silane, trimethyl-2-propynyl, trimethyl 2-propynyl silane, trimethyl prop-2-ynyl silane, trimethyl propargyl silane, silane, trimethyl-2-propyn-1-yl, trimethyl prop-2-yn-1-yl silane, trimethyl-2-propynylsilane, propargyl trimethylsilane, 2-propynyltrimethylsilane | |
| ULYLMHUHFUQKOE-UHFFFAOYSA-N | |
| trimethyl(prop-2-ynyl)silane | |
| 0.1 wt% hydroquinone | |
| 112.25 | |
| Liquid |
| Yellow | |
| 91°C to 93°C | |
| Authentic | |
| Glass bottle | |
| HC∽CCH2Si(CH3)3 | |
| 1 g | |
| 0.75 | |
| Solubility in water: insoluble | |
| C[Si](C)(C)CC#C | |
| 112.25 | |
| 83378 | |
| 80-90% | |
| Propargyltrimethylsilane, 90% |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Danger
EINECSNumber : 236-427-6
Recommended Storage : Freezer -20°C
RUO – Research Use Only