missing translation for 'onlineSavingsMsg'
Learn More

Propargylamine 97.0+%, TCI America™

Catalog No. p-7134841
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P09115ML 5 mL
P091125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P09115ML Supplier TCI America Supplier No. P09115ML
Only null left

Chemical Identifiers

CAS 2450-71-7
Molecular Formula C3H5N
Molecular Weight (g/mol) 55.08
MDL Number MFCD00008198
InChI Key JKANAVGODYYCQF-UHFFFAOYSA-N
Synonym propargylamine, 2-propynylamine, 2-propyn-1-amine, 3-aminopropyne, 3-amino-1-propyne, 2-propyn-1-thiol, 3-aminoprop-1-yne, propargylamin, pro-pargylamine, propargyl amine
PubChem CID 239041
IUPAC Name prop-2-yn-1-amine
SMILES C#CCN

Specifications

Color Yellow
Boiling Point 84°C
Quantity 5 mL
UN Number 3286
Formula Weight 55.08
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Propargylamine
EINECSNumber (2)-2382
RTECSNumber UK5250000
TSCA Yes
Recommended Storage Refrigerator